Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026613.4 | NP_080889.3 | 3 | 620 | ||
GenBank mRNA | AK011768.1 | - | 2 | 136 | ||
AK015873.1 | - | 2 | 457 | |||
AK019247.1 | - | 3 | 591 | |||
AK033555.1 | - | 3 | 599 | |||
AK147056.1 | - | 3 | 621 | |||
BC049229.1 | - | 3 | 353 | |||
BC062248.1 | - | 3 | 566 | |||
BC089458.1 | - | 3 | 458 | |||
BC108339.1 | - | 3 | 603 | |||
BC119347.1 | - | 2 | 165 | 8734096 | ||
BC120580.1 | - | 2 | 165 | 40129503 |