Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024474.2 | NP_001019645.1 | 2 | 919 | ||
GenBank mRNA | AK041534.1 | - | 2 | 1362 | ||
AK136096.1 | - | 2 | 920 | |||
AK136101.1 | - | 2 | 920 | |||
BC086799.1 | - | 2 | 737 | 6833911 |