Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024222.2 | NP_077184.2 | 15 | 2573 | ||
GenBank mRNA | AK012153.1 | - | 15 | 2243 | ||
AK018758.2 | - | 15 | 2283 | |||
AK145674.1 | - | 15 | 2214 | |||
AK148065.1 | - | 15 | 2272 | |||
AK152899.1 | - | 15 | 2621 | |||
AK154979.1 | - | 15 | 2262 | |||
AK170079.1 | - | 15 | 2285 | |||
AX799084.1 | - | 15 | 2284 | |||
BC003206.1 | - | 10 | 1285 | |||
BC052433.1 | - | 15 | 2573 | 6837097 |