Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026697.3 | NP_080973.1 | 7, 8 | 838 | ||
GenBank mRNA | AB232609.1 | - | 6, 7 | 469 | ||
AJ968955.1 | - | 7, 8 | 693 | |||
AJ968956.1 | - | 6, 7 | 639 | |||
AJ968957.1 | - | 7, 8 | 693 | |||
AJ968958.1 | - | 6, 7 | 639 | |||
AJ968959.1 | - | 7, 8 | 693 | |||
AJ968960.1 | - | 6, 7 | 639 | |||
AK002704.1 | - | 7, 8 | 710 | |||
AK133963.1 | - | 7, 8 | 738 | |||
AK135751.1 | - | 7, 8 | 728 | |||
AK160859.1 | - | 7, 8 | 838 | |||
AK165812.1 | - | 7, 8 | 685 | |||
AK168551.1 | - | 7, 8 | 695 | |||
AK168867.1 | - | 7, 8 | 707 | |||
AK169396.1 | - | 7, 8 | 709 | |||
AK181684.1 | - | 4, 5 | 246 | |||
AK216803.1 | - | 4, 5 | 256 | |||
BC009085.1 | - | 7, 8 | 521 | 2649852 | ||
BC025139.1 | - | 7, 8 | 709 | 3980228 | ||
BC056648.1 | - | 7, 8 | 711 | 5034519 |