Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291235.1 | NP_001278164.1 | 4 | 408 | ||
NM_001291236.1 | NP_001278165.1 | 4 | 884 | |||
NM_001291237.1 | NP_001278166.1 | 5 | 592 | |||
NM_026708.2 | NP_080984.1 | 4 | 719 | |||
GenBank mRNA | AK002265.1 | - | 4 | 667 | ||
AK161721.1 | - | 619 | ||||
AK165432.1 | - | 831 | ||||
BC005702.1 | - | 4 | 611 | 3598444 | ||
BC039182.1 | - | 4 | 854 | 3500908 | ||
BG083251.2 | - | 4 | 466 |