Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026718.2 | NP_080994.2 | 13 | 1486 | ||
GenBank mRNA | AK039432.1 | - | 13 | 1681 | ||
AK156539.1 | - | 13 | 1679 | |||
AK162340.1 | - | 5 | 507 | |||
BC027765.1 | - | 11 | 1161 | |||
BC049187.1 | - | 13 | 1665 | |||
BC057884.1 | - | 2 | 77 | |||
BC061692.1 | - | 13 | 1486 | 6308996 | ||
BC108413.1 | - | 13 | 1961 | 6487251 |