Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290765.1 | NP_001277694.1 | 8 | 1042 | ||
NM_026721.3 | NP_080997.1 | 8 | 1003 | |||
GenBank mRNA | AK003191.1 | - | 4 | 305 | ||
AK035296.1 | - | 8 | 910 | |||
BC100463.1 | - | 8 | 1010 | 30918840 |