Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026732.2 | NP_081008.1 | 3 | 389 | ||
GenBank mRNA | AK003500.1 | - | 3 | 356 | ||
AK050273.1 | - | 3 | 375 | |||
BC027021.1 | - | 3 | 389 | 5374870 |