Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002401.3 | NP_002392.2 | 15 | 1950 | ||
NM_203351.1 | NP_976226.1 | 16 | 2043 | |||
XM_005257376.3 | XP_005257433.1 | 16 | 2263 | |||
XM_005257377.3 | XP_005257434.1 | 15 | 1662 | |||
GenBank mRNA | AF172823.1 | - | 1 | 144 | ||
AK225784.1 | - | 16 | 2047 | |||
AK308152.1 | - | 10 | 1158 | |||
AK315305.1 | - | 16 | 1729 | |||
AL834303.1 | - | 16 | 2042 | |||
AX775739.1 | - | 17 | 1790 | |||
AX775741.1 | - | 17 | 1790 | |||
BC008336.1 | - | 14 | 1535 | |||
BC090859.1 | - | 15 | 1951 | 6183752 | ||
BC093672.1 | - | 15 | 1681 | 7939517 | ||
BC093674.1 | - | 15 | 1681 | 7939519 | ||
BC143224.1 | - | 15 | 1669 | |||
BC143235.1 | - | 15 | 1678 | |||
DQ454151.1 | - | 14 | 1619 | |||
U78876.1 | - | 15 | 1714 |