Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163416.1 | NP_001156888.1 | 3 | 1137 | ||
NM_026747.1 | NP_081023.1 | 3 | 1227 | |||
GenBank mRNA | AK003777.1 | - | 2 | 696 | ||
BC107380.2 | - | 3 | 1120 | 40057084 | ||
BC107381.2 | - | 3 | 1210 | 40057088 | ||
BC115902.1 | - | 3 | 765 | |||
BC118508.1 | - | 3 | 1210 | 40106397 |