Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145920.3 | NP_666032.1 | 17 | 3131 | ||
GenBank mRNA | AB083066.1 | - | 17 | 3061 | ||
AK003753.1 | - | 4 | 633 | |||
BC037473.1 | - | 17 | 3112 | 4237472 | ||
BC064473.1 | - | 17 | 3122 | 3156380 |