Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nufip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024205.2 | NP_001019376.1 | 2 | 1661 | ||
GenBank mRNA | AK030113.1 | - | 2 | 1663 | ||
AK122494.1 | - | 2 | 1645 | |||
AK132603.1 | - | 1629 | ||||
AK142954.1 | - | 2 | 1631 | |||
AK152336.1 | - | 2 | 1632 | |||
AK160601.1 | - | 2 | 1662 | |||
AK170764.1 | - | 2 | 1659 |