Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026774.2 | NP_081050.2 | 3, 4 | 434 | ||
GenBank mRNA | AI552736.1 | - | 3, 4 | 434 | ||
AK003358.1 | - | 3, 4 | 274 | |||
AK146289.1 | - | 3, 4 | 294 | |||
BC028547.1 | - | 3, 4 | 264 | 1513442 |