Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024226.4 | NP_077188.1 | 5, 6 | 795 | ||
NM_194051.3 | NP_918940.1 | 7, 8 | 3082 | |||
NM_194052.3 | NP_918941.1 | 5, 6 | 1271 | |||
NM_194053.3 | NP_918942.1 | 6, 7 | 1328 | |||
NM_194054.3 | NP_918943.1 | 7, 8 | 3689 | |||
GenBank mRNA | AB073672.1 | - | 5, 6 | 1647 | ||
AF326337.1 | - | 5, 6 | 575 | |||
AK034902.1 | - | 5, 6 | 1215 | |||
AK206171.1 | - | 2, 3 | 99 | |||
AK220511.1 | - | 7, 8 | 3649 | |||
AY102280.1 | - | 7, 8 | 3082 | |||
AY102281.1 | - | 5, 6 | 1228 | |||
AY102282.1 | - | 6, 7 | 1285 | |||
AY102283.1 | - | 5, 6 | 788 | |||
AY102284.1 | - | 7, 8 | 3646 | |||
AY114152.1 | - | 7, 8 | 3647 | |||
AY114153.1 | - | 6, 7 | 1102 | |||
BC032192.1 | - | 5, 6 | 1941 | 5323152 | ||
BC032272.1 | - | 5, 6 | 2795 | |||
BC056373.1 | - | 7, 8 | 3517 | |||
BU848611.1 | - | 5, 6 | 634 |