Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005924.4 | NP_005915.2 | 1 | 377 | ||
GenBank mRNA | AK313386.1 | - | 1 | 305 | ||
BC017021.1 | - | 1 | 277 | 3917118 | ||
CR536553.1 | - | 1 | 96 | |||
DA586635.1 | - | 1 | 377 | |||
L36328.1 | - | 1 | 128 | |||
X82629.1 | - | 1 | 361 |