Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam172a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163419.1 | NP_001156891.1 | 2 | 185 | ||
NM_001163420.1 | NP_001156892.1 | 2 | 185 | |||
NM_138312.1 | NP_612185.1 | 3 | 309 | |||
GenBank mRNA | AF064782.1 | - | 2 | 138 | ||
AK049727.1 | - | 2 | 146 | |||
AK052419.1 | - | 2 | 157 | |||
AK165273.1 | - | 2 | 159 | |||
AK166285.1 | - | 2 | 157 | |||
BC057380.1 | - | 2 | 153 |