Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199186.1 | NP_001186115.1 | 5 | 357 | ||
NM_152808.3 | NP_690021.1 | 5 | 359 | |||
GenBank mRNA | AK029457.1 | - | 5 | 319 | ||
AK041533.1 | - | 5 | 316 | |||
AK043924.1 | - | 5 | 329 | |||
AK048648.1 | - | 5 | 344 | |||
AK088553.1 | - | 5 | 306 | |||
AK153992.1 | - | 5 | 338 | |||
AK166695.1 | - | 5 | 360 | |||
BC023763.1 | - | 5 | 342 | |||
BC031535.1 | - | 5 | 335 | 3991609 |