Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199186.1 | NP_001186115.1 | 6 | 488 | ||
NM_152808.3 | NP_690021.1 | 6 | 490 | |||
GenBank mRNA | AK029457.1 | - | 6 | 450 | ||
AK041533.1 | - | 6 | 447 | |||
AK043924.1 | - | 6 | 460 | |||
AK048648.1 | - | 6 | 475 | |||
AK088553.1 | - | 6 | 437 | |||
AK153992.1 | - | 6 | 469 | |||
AK166695.1 | - | 6 | 491 | |||
AK186248.1 | - | 1 | 38 | |||
BC023763.1 | - | 6 | 473 | |||
BC031535.1 | - | 6 | 466 | 3991609 |