Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026816.3 | NP_081092.1 | 6, 7 | 602 | ||
GenBank mRNA | AK144858.1 | - | 6, 7 | 603 | ||
BC027173.1 | - | 6, 7 | 598 | 3986466 |