Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifitm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112715.1 | NP_001106186.1 | 1 | 259 | ||
NM_026820.3 | NP_081096.3 | 2 | 233 | |||
GenBank mRNA | AK004121.1 | - | 1 | 122 | ||
AK169960.1 | - | 1 | 104 | |||
BC027285.1 | - | 2 | 449 | 3492212 | ||
BC090258.1 | - | 1 | 87 | 4982747 | ||
BC090972.1 | - | 1 | 187 | 30297959 | ||
BY198144.1 | - | 1 | 259 |