Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080575.2 | NP_542142.1 | 12 | 1779 | ||
GenBank mRNA | AB046742.1 | - | 12 | 1768 | ||
AK088244.1 | - | 12 | 1758 | |||
AK146568.1 | - | 12 | 1780 | |||
AK158119.1 | - | 12 | 1774 | |||
AK172412.1 | - | 12 | 1765 | |||
AK172470.1 | - | 12 | 1754 | |||
AK173275.1 | - | 12 | 1740 | |||
BC030930.1 | - | 8 | 953 | 4911679 | ||
BC092278.1 | - | 12 | 1808 | 3710505 |