Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080575.2 | NP_542142.1 | 10, 11 | 1598 | ||
GenBank mRNA | AB046742.1 | - | 10, 11 | 1587 | ||
AK088244.1 | - | 10, 11 | 1577 | |||
AK146568.1 | - | 10, 11 | 1599 | |||
AK158119.1 | - | 10, 11 | 1593 | |||
AK172470.1 | - | 10, 11 | 1573 | |||
AK173275.1 | - | 10, 11 | 1559 | |||
BC030930.1 | - | 6, 7 | 772 | 4911679 | ||
BC092278.1 | - | 10, 11 | 1627 | 3710505 |