Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175134.4 | NP_780343.1 | 5 | 834 | ||
GenBank mRNA | AB076382.1 | - | 5 | 829 | ||
AK079270.1 | - | 3 | 393 | |||
AK088304.1 | - | 5 | 795 | |||
AK148049.1 | - | 5 | 795 | |||
BC035553.1 | - | 5 | 2412 | |||
BC086659.1 | - | 5 | 801 | 30607272 | ||
BC094595.1 | - | 5 | 797 | 5707829 |