Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164224.1 | NP_001157696.1 | 9 | 1049 | ||
NM_026877.2 | NP_081153.1 | 10 | 1270 | |||
NM_198223.2 | NP_937866.1 | 9 | 1049 | |||
GenBank mRNA | AK004509.1 | - | 4 | 282 | ||
AK088197.1 | - | 10 | 1108 | |||
AK146061.1 | - | 9 | 1050 | |||
AY349132.1 | - | 8 | 1039 | |||
AY349133.1 | - | 10 | 1187 | |||
BC019177.1 | - | 10 | 1213 | 5008127 | ||
BC022115.1 | - | 10 | 1217 | 5101238 | ||
BC031153.1 | - | 10 | 1202 | 4976569 |