Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026879.2 | NP_081155.1 | 5 | 677 | ||
GenBank mRNA | AK004506.1 | - | 2 | 150 | ||
AK084111.1 | - | 5 | 679 | |||
AK210184.1 | - | 2 | 126 | |||
AK216188.1 | - | 2 | 126 | |||
AK216245.1 | - | 2 | 126 | |||
AK217551.1 | - | 2 | 126 | |||
BC042626.1 | - | 4 | 433 | |||
BC055809.1 | - | 5 | 706 | 6414077 |