Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039483.1 | NP_001034572.1 | 3 | 360 | ||
GenBank mRNA | AK004504.1 | - | 3 | 202 | ||
AK015068.1 | - | 3 | 651 | |||
AK037709.1 | - | 3 | 295 | |||
AK162344.1 | - | 3 | 299 | |||
BC011457.1 | - | 3 | 234 | 3585288 | ||
BC019761.1 | - | 3 | 278 | 5009960 | ||
BC020098.1 | - | 3 | 290 | 4019484 |