Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026892.3 | NP_081168.1 | 3 | 435 | ||
GenBank mRNA | AK005192.1 | - | 3 | 402 | ||
AK013965.1 | - | 3 | 408 | |||
BC030319.1 | - | 3 | 304 | 5372299 | ||
BC033505.1 | - | 3 | 406 | 4209443 |