Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163421.1 | NP_001156893.1 | 7 | 538 | ||
NM_001163422.1 | NP_001156894.1 | 7 | 538 | |||
NM_026895.1 | NP_081171.1 | 7 | 538 | |||
GenBank mRNA | AK005193.1 | - | 7 | 509 | ||
AK156071.1 | - | 7 | 489 | |||
BC061248.1 | - | 7 | 510 | |||
BC094396.1 | - | 2 | 71 |