Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haghl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271433.1 | NP_001258362.1 | 7 | 781 | ||
NM_001271434.1 | NP_001258363.1 | 7 | 781 | |||
NM_001271435.1 | NP_001258364.1 | 7 | 781 | |||
NM_001271436.1 | NP_001258365.1 | 6 | 595 | |||
NM_001271437.1 | NP_001258366.1 | 6 | 590 | |||
NM_026897.3 | NP_081173.1 | 6 | 766 | |||
GenBank mRNA | AK005274.1 | - | 6 | 762 | ||
AK012748.1 | - | 7 | 778 | |||
AK021220.1 | - | 6 | 597 | |||
AK032999.1 | - | 4 | 739 | |||
AK034798.1 | - | 7 | 756 | |||
AK047517.1 | - | 7 | 736 | |||
AK079632.1 | - | 4 | 930 | |||
AK080454.1 | - | 1 | 1717 | |||
AK154248.1 | - | 6 | 571 | |||
BC005586.1 | - | 7 | 727 | |||
BC030466.1 | - | 7 | 766 | 5253213 | ||
BC083322.1 | - | 7 | 720 | 6820885 |