Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133706.2 | NP_598467.1 | 2 | 226 | ||
GenBank mRNA | AK155107.1 | - | 2 | 238 | ||
AK171954.1 | - | 2 | 205 | |||
BC018156.1 | - | 2 | 226 | 4192256 | ||
BC093511.1 | - | 2 | 203 | 5124076 | ||
X81718.1 | - | 1 | 18 |