Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026934.3 | NP_081210.2 | 4 | 413 | ||
GenBank mRNA | AK005661.1 | - | 4 | 511 | ||
AK012012.1 | - | 4 | 503 | |||
AK139511.1 | - | 4 | 508 | |||
AK152374.1 | - | 4 | 504 | |||
AK159596.1 | - | 4 | 485 | |||
AK167696.1 | - | 4 | 506 | |||
BC031845.1 | - | 4 | 413 | 3983096 | ||
BC058229.1 | - | 4 | 340 | 6313954 |