Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026936.3 | NP_081212.1 | 10 | 1251 | ||
GenBank mRNA | AK007567.1 | - | 10 | 1253 | ||
AK028379.1 | - | 10 | 1250 | |||
AK044817.1 | - | 10 | 1250 | |||
AK052432.1 | - | 10 | 1252 | |||
AK077624.1 | - | 10 | 1252 | |||
AK087756.1 | - | 10 | 1255 | |||
AK139484.1 | - | 10 | 1259 | |||
AK154304.1 | - | 10 | 1254 | |||
AK154532.1 | - | 10 | 1252 | |||
AK170898.1 | - | 10 | 1252 | |||
BC027191.1 | - | 11 | 1183 | 4223847 |