Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026955.2 | NP_081231.1 | 3 | 725 | ||
GenBank mRNA | AK008621.1 | - | 3 | 726 | ||
AK034938.1 | - | 3 | 726 | |||
AK161045.1 | - | 3 | 518 | |||
BC055786.1 | - | 3 | 687 | 6404797 |