Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199188.1 | NP_001186117.1 | 13 | 1353 | ||
NM_024225.5 | NP_077187.1 | 12 | 1264 | |||
GenBank mRNA | AK007676.1 | - | 12 | 1212 | ||
AK029463.1 | - | 12 | 1224 | |||
AK031873.1 | - | 13 | 1245 | |||
AK049129.1 | - | 13 | 1245 | |||
AK078776.1 | - | 13 | 1246 | |||
AK144710.1 | - | 13 | 1249 | |||
AK152311.1 | - | 13 | 1290 | |||
AK159115.1 | - | 9 | 807 | |||
AK159904.1 | - | 12 | 1169 | |||
AK167362.1 | - | 12 | 1184 | |||
AK169545.1 | - | 11 | 2159 | |||
AK169786.1 | - | 13 | 1245 | |||
AK217080.1 | - | 2 | 132 | |||
AK219351.1 | - | 2 | 133 | |||
BC002242.1 | - | 13 | 1322 | 3492262 |