Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027001.1 | NP_081277.1 | 3 | 1100 | ||
GenBank mRNA | BC033350.1 | - | 3 | 1100 | 5362778 | |
BC034642.1 | - | 3 | 1060 | 5363521 |