Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027016.2 | NP_081292.1 | 2 | 565 | ||
GenBank mRNA | AK013922.1 | - | 2 | 380 | ||
AK088093.1 | - | 2 | 114 | |||
AK090078.1 | - | 2 | 115 | |||
BC017162.1 | - | 2 | 566 | |||
BC048532.1 | - | 2 | 115 | |||
BC053504.1 | - | 2 | 102 | |||
BC067202.1 | - | 2 | 100 | 6332701 | ||
BC071243.1 | - | 2 | 115 |