Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000247.2 | NP_000238.1 | 5 | 1058 | ||
NM_001177519.2 | NP_001170990.1 | 5 | 1062 | |||
NM_001289152.1 | NP_001276081.1 | 5 | 1926 | |||
NM_001289153.1 | NP_001276082.1 | 5 | 1906 | |||
NM_001289154.1 | NP_001276083.1 | 5 | 1734 | |||
GenBank mRNA | AB506764.1 | - | 5 | 992 | ||
AK225393.1 | - | 5 | 1046 | |||
AK299135.1 | - | 5 | 1034 | |||
AY204547.1 | - | 5 | 995 | |||
AY750850.1 | - | 5 | 992 | |||
BC016929.1 | - | 5 | 1046 | 4446575 | ||
L14848.1 | - | 5 | 1031 |