Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MID1 SIRNA Assays ›
Gene Symbol:
MID1
Gene Name:
midline 1
Gene Aliases:
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
Chromosome Location:
Chr.X: 10445310 - 11111264 on Build GRCh38
UniGene ID:
Hs.27695
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8778
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MID1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000381.3
NP_000372.1
4
1239
NM_001098624.2
NP_001092094.1
4
1169
NM_001193277.1
NP_001180206.1
4
1028
NM_001193278.1
NP_001180207.1
4
1047
NM_001193279.1
NP_001180208.1
3
894
NM_001193280.1
NP_001180209.1
3
780
NM_033289.1
NP_150631.1
4
1125
NM_033290.3
NP_150632.1
4
1031
XM_005274536.1
XP_005274593.1
5
1204
XM_005274537.1
XP_005274594.1
5
1181
XM_006724492.3
XP_006724555.1
5
1803
XM_011545525.2
XP_011543827.1
7
1505
XM_011545526.2
XP_011543828.1
7
1540
XM_011545527.2
XP_011543829.1
5
1450
XM_017029534.1
XP_016885023.1
5
1370
XM_017029535.1
XP_016885024.1
7
1391
XM_017029536.1
XP_016885025.1
6
1467
XM_017029537.1
XP_016885026.1
6
1238
XM_017029538.1
XP_016885027.1
4
937
XM_017029539.1
XP_016885028.1
4
1099
XM_017029540.1
XP_016885029.1
6
1352
XM_017029541.1
XP_016885030.1
7
1505
XM_017029542.1
XP_016885031.1
6
1352
GenBank mRNA
AF035360.1
-
4
925
AF041206.1
-
4
1119
AF041208.1
-
4
1239
AF041210.1
-
3
728
AF230976.1
-
4
1024
AF230977.1
-
4
1024
AF269101.1
-
5
1147
AK025187.1
-
3
210
AK226104.1
-
4
943
AK291215.1
-
4
1121
AK297205.1
-
5
1035
AK299061.1
-
4
1064
AK315095.1
-
4
1070
AY539962.1
-
3
221
AY539963.1
-
4
382
AY539964.1
-
4
374
AY539979.1
-
4
325
AY539980.1
-
3
255
AY539981.1
-
2
102
AY539982.1
-
2
110
AY539983.1
-
3
264
AY539984.1
-
3
264
AY539985.1
-
3
464
AY539986.1
-
2
311
AY539994.1
-
4
991
AY539995.1
-
4
606
AY539996.1
-
4
492
AY539997.1
-
2
138
AY539998.1
-
3
331
AY540014.1
-
3
339
AY540015.1
-
3
453
AY540022.1
-
3
838
AY540023.1
-
3
724
BC053626.1
-
4
1096
6163881
Y13667.1
-
4
1024
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
negative regulation of microtubule depolymerization
pattern specification process
positive regulation of stress-activated MAPK cascade
protein localization to microtubule
interferon-gamma-mediated signaling pathway
protein binding
microtubule binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0