Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033149.2 | NP_001028321.1 | 1 | 239 | ||
GenBank mRNA | AK133456.1 | - | 1 | 240 | ||
AK161499.1 | - | 1 | 39 | |||
AK172918.1 | - | 359 | ||||
BC028891.1 | - | 1 | 323 | |||
BC139242.1 | - | 1 | 237 | 8862264 | ||
BC139244.1 | - | 1 | 237 | 8862266 |