Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATXN3 SIRNA Assays ›
Gene Symbol:
ATXN3
Gene Name:
ataxin 3
Gene Aliases:
AT3, ATX3, JOS, MJD, MJD1, SCA3
Chromosome Location:
Chr.14: 92058552 - 92106621 on Build GRCh38
UniGene ID:
Hs.532632
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8795
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127696.1
NP_001121168.1
3
300
NM_001164778.1
NP_001158250.1
4
345
NM_001164781.1
NP_001158253.1
2
135
NM_004993.5
NP_004984.2
4
345
NM_030660.4
NP_109376.1
3
180
NR_028459.1
4
345
NR_028461.1
4
345
GenBank mRNA
AB050194.1
-
3
169
AK225884.1
-
4
284
AK314919.1
-
4
297
BC033711.1
-
4
288
4393766
BC095402.1
-
4
305
30347215
BU190081.1
-
3
279
DA827537.1
-
4
345
GQ176433.1
-
4
298
GQ176434.1
-
4
297
GQ176435.1
-
4
298
GQ176440.1
-
3
133
GQ176448.1
-
4
298
GQ176452.1
-
3
298
GQ176455.1
-
3
133
GQ176462.1
-
4
298
GQ176467.1
-
3
133
GQ176476.1
-
4
298
GQ176486.1
-
3
253
GQ176488.1
-
4
298
GQ176489.1
-
3
253
GQ176494.1
-
4
297
GQ176498.1
-
3
133
GQ176499.1
-
4
298
GQ176504.1
-
4
298
GQ176508.1
-
4
298
GQ176510.1
-
4
298
GQ176512.1
-
4
295
GQ176516.1
-
4
298
GQ176523.1
-
4
298
GQ176526.1
-
2
88
GQ176532.1
-
3
298
GQ176533.1
-
4
298
GQ176536.1
-
3
133
GQ176539.1
-
3
133
GQ176548.1
-
4
298
GQ176550.1
-
4
298
GQ176551.1
-
4
297
GQ176552.1
-
4
297
GQ176560.1
-
4
298
GQ176563.1
-
4
298
GQ176565.1
-
4
298
GQ176566.1
-
3
133
GQ176567.1
-
4
298
GQ176568.1
-
4
298
GQ176570.1
-
4
298
GQ176573.1
-
3
133
GQ176575.1
-
4
298
GQ176576.1
-
4
298
GQ176577.1
-
4
298
GQ176578.1
-
4
298
GQ176580.1
-
4
298
GQ176581.1
-
298
GQ176589.1
-
4
298
GQ176592.1
-
3
133
GQ176593.1
-
4
297
GQ176594.1
-
4
298
GQ176599.1
-
4
298
GQ176602.1
-
4
316
GQ176609.1
-
298
GQ176612.1
-
4
298
GQ176615.1
-
298
GQ176616.1
-
4
298
GQ176618.1
-
124
GQ176619.1
-
4
298
GQ176621.1
-
4
298
GQ176622.1
-
4
298
GQ176624.1
-
4
298
GQ176628.1
-
298
GQ176632.1
-
4
298
GQ176640.1
-
4
298
GQ176642.1
-
4
298
GQ176644.1
-
4
293
GQ176645.1
-
4
298
GQ176646.1
-
4
298
GQ176658.1
-
4
298
GQ176660.1
-
3
212
GQ176665.1
-
3
133
GQ176668.1
-
4
295
GQ176671.1
-
298
GQ176673.1
-
4
298
GQ176674.1
-
4
298
GQ176675.1
-
3
133
GQ176677.1
-
4
298
GQ176679.1
-
3
133
GQ176684.1
-
4
298
GQ176685.1
-
4
298
GQ176686.1
-
298
GQ176687.1
-
4
298
GQ176692.1
-
4
298
GQ176694.1
-
3
133
GQ176697.1
-
4
298
GQ176698.1
-
298
GQ176699.1
-
298
GQ176700.1
-
3
133
GQ176701.1
-
4
298
GQ176703.1
-
4
298
GQ176704.1
-
4
298
GQ176708.1
-
4
298
GQ176712.1
-
3
253
GQ176714.1
-
4
298
GQ176715.1
-
3
133
GQ176721.1
-
4
298
GQ176725.1
-
4
298
GQ176728.1
-
4
298
GQ176735.1
-
4
298
GQ176737.1
-
4
298
GQ176741.1
-
298
GQ176744.1
-
3
253
GQ176745.1
-
298
GQ176746.1
-
4
298
GQ176747.1
-
298
GQ176749.1
-
4
298
GQ176750.1
-
3
133
GQ176752.1
-
298
GQ176754.1
-
3
133
GQ176756.1
-
4
298
GQ176759.1
-
4
298
GQ176762.1
-
298
GQ176763.1
-
4
298
GQ176766.1
-
4
298
GQ176771.1
-
4
298
GQ176776.1
-
298
GQ176787.1
-
4
298
GQ176791.1
-
4
298
GQ176800.1
-
298
GQ176805.1
-
4
274
GQ176806.1
-
4
297
GQ176808.1
-
298
GQ176813.1
-
3
133
GQ176820.1
-
4
298
GQ176822.1
-
3
133
GQ176824.1
-
4
298
GQ176825.1
-
298
GQ176826.1
-
4
298
GQ176827.1
-
4
298
GQ176829.1
-
4
298
GQ176830.1
-
269
GQ176831.1
-
298
GQ176835.1
-
3
132
GQ176836.1
-
3
133
GQ176837.1
-
4
298
GQ176838.1
-
4
298
S75313.1
-
4
311
U64820.1
-
4
333
U64821.1
-
4
296
U64822.1
-
3
273
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
nucleotide-excision repair
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
misfolded or incompletely synthesized protein catabolic process
chemical synaptic transmission
nervous system development
regulation of cell-substrate adhesion
protein deubiquitination
actin cytoskeleton organization
cellular response to heat
monoubiquitinated protein deubiquitination
exploration behavior
proteasome-mediated ubiquitin-dependent protein catabolic process
intermediate filament cytoskeleton organization
protein K63-linked deubiquitination
histone H3 deacetylation
protein K48-linked deubiquitination
cellular response to misfolded protein
positive regulation of ERAD pathway
protein localization to cytosolic proteasome complex involved in ERAD pathway
RNA polymerase II regulatory region DNA binding
histone deacetylase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
omega peptidase activity
ubiquitin protein ligase binding
identical protein binding
ATPase binding
Lys63-specific deubiquitinase activity
Lys48-specific deubiquitinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0