Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190453.1 | NP_001177382.1 | 15 | 1381 | ||
NM_001190454.1 | NP_001177383.1 | 14 | 1390 | |||
NM_027151.2 | NP_081427.1 | 14 | 1375 | |||
GenBank mRNA | AK009749.1 | - | 9 | 628 | ||
AK144464.1 | - | 14 | 1251 | |||
AK144505.1 | - | 14 | 1252 | |||
AK164021.1 | - | 14 | 1252 | |||
AK180539.1 | - | 5 | 349 | |||
BC004613.1 | - | 14 | 1238 | 3585310 |