Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mif4gd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243584.1 | NP_001230513.1 | 5 | 617 | ||
NM_027162.4 | NP_081438.4 | 4 | 901 | |||
GenBank mRNA | AK003708.1 | - | 678 | |||
AK010175.1 | - | 5 | 571 | |||
AK150749.1 | - | 5 | 571 | |||
BC019515.1 | - | 5 | 528 | 4460171 | ||
BC026740.1 | - | 5 | 577 | 4015105 | ||
BC055812.1 | - | 4 | 901 | 4934728 |