Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130410.1 | NP_001123882.1 | 8 | 955 | ||
NM_001607.3 | NP_001598.1 | 10 | 1234 | |||
NR_024024.1 | 8 | 1359 | ||||
XM_006713122.1 | XP_006713185.1 | 9 | 939 | |||
XM_006713123.2 | XP_006713186.1 | 9 | 1076 | |||
XM_011533650.2 | XP_011531952.1 | 8 | 1102 | |||
GenBank mRNA | AF035295.1 | - | 8 | 1090 | ||
AK025138.1 | - | 10 | 1068 | |||
AK127051.1 | - | 5 | 1699 | |||
AK293716.1 | - | 11 | 1023 | |||
BC000635.2 | - | 10 | 1121 | 3345271 | ||
BC011977.1 | - | 10 | 1069 | 3860150 | ||
BC014474.1 | - | 8 | 753 | 4908159 | ||
X12966.1 | - | 10 | 1033 | |||
X14813.1 | - | 10 | 1063 | |||
X51460.1 | - | 8 | 735 |