Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023529.2 | NP_076018.2 | 6 | 579 | ||
GenBank mRNA | AB026046.1 | - | 6 | 588 | ||
AF237918.1 | - | 6 | 579 | |||
AK008019.1 | - | 4 | 438 | |||
BC144935.1 | - | 6 | 576 | 9053483 | ||
BC150675.1 | - | 6 | 576 | 9087586 |