Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039394.1 | NP_001034483.1 | 2 | 572 | ||
NM_133717.3 | NP_598478.1 | 2 | 278 | |||
GenBank mRNA | AB232640.1 | - | 2 | 337 | ||
AJ534465.1 | - | 2 | 556 | |||
AK007834.1 | - | 2 | 168 | |||
AK010874.1 | - | 2 | 571 | |||
AK158283.1 | - | 2 | 573 | |||
BC010248.1 | - | 2 | 278 | 3155089 | ||
BY151021.1 | - | 2 | 237 | |||
BY707837.1 | - | 2 | 165 | |||
BY753078.1 | - | 2 | 573 | |||
CV782943.1 | - | 2 | 178 |