Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172402.3 | NP_765990.2 | 4 | 641 | ||
GenBank mRNA | AK031187.1 | - | 4 | 446 | ||
AK145178.1 | - | 4 | 601 | |||
AK167501.1 | - | 4 | 451 | |||
AK170643.1 | - | 4 | 610 | |||
AK172029.1 | - | 4 | 609 | |||
AK172597.1 | - | 4 | 599 | |||
BB641480.1 | - | 4 | 642 | |||
BC031874.1 | - | 4 | 575 | 5149984 |