Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027268.2 | NP_081544.1 | 4 | 563 | ||
GenBank mRNA | AK012765.1 | - | 4 | 591 | ||
AK032573.1 | - | 4 | 590 | |||
AK050481.1 | - | 4 | 581 | |||
AK082953.1 | - | 4 | 563 | |||
AK089968.1 | - | 4 | 553 | |||
AK129084.1 | - | 4 | 550 | |||
AK145904.1 | - | 4 | 565 | |||
BC023889.1 | - | 4 | 550 | 5252046 | ||
EF410099.1 | - | 514 |