Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133718.4 | NP_598479.1 | 2 | 542 | ||
GenBank mRNA | AK008408.1 | - | 2 | 428 | ||
AK077065.1 | - | 2 | 545 | |||
AK150071.1 | - | 2 | 445 | |||
AK159364.1 | - | 2 | 469 | |||
AK161096.1 | - | 2 | 424 | |||
AK170171.1 | - | 2 | 445 | |||
AK170697.1 | - | 2 | 446 | |||
AK178013.1 | - | 2 | 182 | |||
AK192822.1 | - | 2 | 182 | |||
AK198710.1 | - | 2 | 183 | |||
AK200429.1 | - | 2 | 206 | |||
AK203327.1 | - | 2 | 183 | |||
BC018367.1 | - | 2 | 394 | 2648510 | ||
BC018491.1 | - | 2 | 422 | 2651602 |