Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dopey2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026700.2 | NP_080976.1 | 2 | 242 | ||
NM_027293.1 | NP_081569.1 | 2 | 242 | |||
GenBank mRNA | AK002808.1 | - | 2 | 172 | ||
AK147255.1 | - | 2 | 220 | |||
AK156170.1 | - | 2 | 216 | |||
AK164839.1 | - | 2 | 219 | |||
BY184348.1 | - | 2 | 242 | |||
BY195296.1 | - | 2 | 242 |