Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027303.1 | NP_081579.1 | 3 | 520 | ||
GenBank mRNA | AK006546.1 | - | 3 | 462 |